Order Entry
Singapore
ContactUsLinkComponent
FLUORESCEIN-CONJUGATED GOAT F(ab')2 FRAGMENT TO HUMAN IgG Fc. Anti-Human Host: goat Form: lyophilized (F(ab)2 ) Applications: Imm unostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. FITC Conjugated 1 * 2 mL
Catalog # ICNA0855184
Supplier:  MP Biomedicals
FLUORESCEIN-CONJUGATED GOAT F(ab')2 FRAGMENT TO HUMAN IgG Fc. Anti-Human Host: goat Form: lyophilized (F(ab)2 ) Applications: Imm unostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. FITC Conjugated 1 * 2 mL
Catalog # ICNA0855184
Supplier:  MP Biomedicals