Order Entry
Singapore
ContactUsLinkComponent
GOAT ANTISERUM TO HUMAN IgD ( D CHAIN). Anti-Human Host: goat Form: lyophilized Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mL
Catalog # ICNA0855083
Supplier:  MP Biomedicals
GOAT ANTISERUM TO HUMAN IgD ( D CHAIN). Anti-Human Host: goat Form: lyophilized Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mL
Catalog # ICNA0855083
Supplier:  MP Biomedicals