Order Entry
Singapore
ContactUsLinkComponent
GOAT IgG FRACTION TO HUMAN SECRETORY IgA. Anti-HumanHost: goatForm: lyophilized (IgG)Applications: Immunostaining of acetone-fixed f rozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 5 mL
Catalog # ICNA0855001
Supplier:  MP Biomedicals
GOAT IgG FRACTION TO HUMAN SECRETORY IgA. Anti-HumanHost: goatForm: lyophilized (IgG)Applications: Immunostaining of acetone-fixed f rozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 5 mL
Catalog # ICNA0855001
Supplier:  MP Biomedicals