Order Entry
Singapore
ContactUsLinkComponent
GOAT ANTISERUM TO HORSERADISH PEROXIDASE. Anti-HRPHost: goatForm: lyophilizedApplications: Immunostaining of acetone-fixed frozen an d formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mL
Catalog # ICNA0855970
Supplier:  MP Biomedicals
GOAT ANTISERUM TO HORSERADISH PEROXIDASE. Anti-HRPHost: goatForm: lyophilizedApplications: Immunostaining of acetone-fixed frozen an d formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mL
Catalog # ICNA0855970
Supplier:  MP Biomedicals