Order Entry
Singapore
ContactUsLinkComponent
ALKALINE PHOSPHATASE - CONJUGATED GOAT AFFINITY PURIFIED ANTIBODY TO HUMAN IMMUNOGLOBULINS (IgG, IgA, IgM). (Anti-IgG, IgA, and IgM) Anti-Human Host: goat Form: affinity purified liquid Applications: Immunostaining of acetone-fixed frozen and formal 1 * 1 mL
Catalog # ICNA0859284
Supplier:  MP Biomedicals
ALKALINE PHOSPHATASE - CONJUGATED GOAT AFFINITY PURIFIED ANTIBODY TO HUMAN IMMUNOGLOBULINS (IgG, IgA, IgM). (Anti-IgG, IgA, and IgM) Anti-Human Host: goat Form: affinity purified liquid Applications: Immunostaining of acetone-fixed frozen and formal 1 * 1 mL
Catalog # ICNA0859284
Supplier:  MP Biomedicals