Order Entry
Singapore
ContactUsLinkComponent
ALKALINE PHOSPHATASE-CONJUGATED GOAT AFFINITY PURIFIED ANTIBODY TO RABBIT IgG Fc. Anti-RabbitHost: goatForm: affinity purified liqui dApplications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections.AP Conjug 1 * 1 mL
  ICNA0859652
ALKALINE PHOSPHATASE-CONJUGATED GOAT AFFINITY PURIFIED ANTIBODY TO RABBIT IgG Fc. Anti-RabbitHost: goatForm: affinity purified liqui dApplications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections.AP Conjug 1 * 1 mL
  ICNA0859652