Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BCIP/NBT PLUS LIQUID SUBSTRATE 1 * 100 mL
Catalog # ICNA08980771
Supplier:
MP Biomedicals
Expand image
BCIP/NBT PLUS LIQUID SUBSTRATE 1 * 100 mL
Catalog # ICNA08980771
Supplier:
MP Biomedicals