$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
TRIS - BORATE - EDTA BUFFER,10X CONCENTRATED SOLUTION. 1 * 4 L
ICNA04816724
:
MP Biomedicals
TRIS - BORATE - EDTA BUFFER,10X CONCENTRATED SOLUTION. 1 * 4 L
ICNA04816724
:
MP Biomedicals