Order Entry
Singapore
ContactUsLinkComponent
FITC CONJUGATED GOAT AFFINITY PURIFIED IgG TO CAT IgG (WHOLE MOLECULE). Anti-CatHost: goatForm: affinity purified liquidApplications : Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections.FITC Conjugated 1 * 2 mg
Catalog # ICNA0856995
Supplier:  MP Biomedicals
FITC CONJUGATED GOAT AFFINITY PURIFIED IgG TO CAT IgG (WHOLE MOLECULE). Anti-CatHost: goatForm: affinity purified liquidApplications : Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections.FITC Conjugated 1 * 2 mg
Catalog # ICNA0856995
Supplier:  MP Biomedicals