Order Entry
Singapore
ContactUsLinkComponent
PURIFIED MOUSE MONOCLONAL ANTIBODY (HIV.OT 39B) TO HUMAN IMMUNODEFICIENCY VIRUS-1 (HIV-1) P24. Anti-HIV-1 p24Clone: OT39BIsotype: mo use IgG1Applications: ELISA Immunoblotting Immunostaining.The antiserum is produced by hybridoma cultured in protei 1 * 100 µG
  ICNA0856980
PURIFIED MOUSE MONOCLONAL ANTIBODY (HIV.OT 39B) TO HUMAN IMMUNODEFICIENCY VIRUS-1 (HIV-1) P24. Anti-HIV-1 p24Clone: OT39BIsotype: mo use IgG1Applications: ELISA Immunoblotting Immunostaining.The antiserum is produced by hybridoma cultured in protei 1 * 100 µG
  ICNA0856980