Order Entry
Singapore
ContactUsLinkComponent
From Streptomyces sp.Antibiotic Inhibits many biochemical functions such as synthesis of glycoproteins, procollagen and peptidoglycans and IgA and IgE secretion by plasma cells. Also affects the fibroblast interferon-cell system. 1 * 5 mg
Catalog # ICNA0215002805
Supplier:  MP Biomedicals
From Streptomyces sp.Antibiotic Inhibits many biochemical functions such as synthesis of glycoproteins, procollagen and peptidoglycans and IgA and IgE secretion by plasma cells. Also affects the fibroblast interferon-cell system. 1 * 5 mg
Catalog # ICNA0215002805
Supplier:  MP Biomedicals