Order Entry
Singapore
ContactUsLinkComponent
ALCIAN BLUE 8 GX, CERTIFIED. C.I. 74240 (Alcian Blue) Useful as a bacterial stain, and as a dye for histiocytes and fibroblasts. 1 * 5 g
Catalog # ICNA0215262405
Supplier:  MP Biomedicals
ALCIAN BLUE 8 GX, CERTIFIED. C.I. 74240 (Alcian Blue) Useful as a bacterial stain, and as a dye for histiocytes and fibroblasts. 1 * 5 g
Catalog # ICNA0215262405
Supplier:  MP Biomedicals