Order Entry
Singapore
ContactUsLinkComponent
Ecolite(+) Liquid Scintillation Fluid. Environmentally Safe, Biodegradable, Non-toxic, and Non-flammable Ready-to-use for a wi de variety of aqueous sample applications where sample load capacity and enhanced tritium counting efficiency is esse 1 * 15,14 L
Catalog # ICNA0188247504
Supplier:  MP Biomedicals
undefined
Ecolite(+) Liquid Scintillation Fluid. Environmentally Safe, Biodegradable, Non-toxic, and Non-flammable Ready-to-use for a wi de variety of aqueous sample applications where sample load capacity and enhanced tritium counting efficiency is esse 1 * 15,14 L
Catalog # ICNA0188247504
Supplier:  MP Biomedicals