Order Entry
Singapore
ContactUsLinkComponent
Z-Asp-Glu-Val-Asp-7-Amino-4-trifluoromethylcoumarin. A fluorogenic substrate for Caspase-3 (CPP32, YAMA). Also acts as a substrate f or granzyme B-processed ICE-LAP3. 1 * 10 mg
Catalog # ICNA03AFC138Z10
Supplier:  MP Biomedicals
Z-Asp-Glu-Val-Asp-7-Amino-4-trifluoromethylcoumarin. A fluorogenic substrate for Caspase-3 (CPP32, YAMA). Also acts as a substrate f or granzyme B-processed ICE-LAP3. 1 * 10 mg
Catalog # ICNA03AFC138Z10
Supplier:  MP Biomedicals