Order Entry
Singapore
ContactUsLinkComponent
A fungal metabolite which potently and selectively inhibits phosphatidylinositol 3-kinase. It also inhibits fMLP induced PIP3 and superoxide anion production in guinea pig neutrophils. It blocks insulin metabolic effects in rat adipocytes without aff 1 * 5 mg
Catalog # ICNA0219569005
Supplier:  MP Biomedicals
A fungal metabolite which potently and selectively inhibits phosphatidylinositol 3-kinase. It also inhibits fMLP induced PIP3 and superoxide anion production in guinea pig neutrophils. It blocks insulin metabolic effects in rat adipocytes without aff 1 * 5 mg
Catalog # ICNA0219569005
Supplier:  MP Biomedicals