Order Entry
Singapore
ContactUsLinkComponent
N - LAUROYLSARCOSINE SODIUM SALT. Molecular Biology Reagent.Sodium SaltPurity: =95.5%Useful in concentrated salt solutions used in the cell lysis step during RNA purification. 1 * 250 g
Catalog # ICNA0219400883
Supplier:  MP Biomedicals
N - LAUROYLSARCOSINE SODIUM SALT. Molecular Biology Reagent.Sodium SaltPurity: =95.5%Useful in concentrated salt solutions used in the cell lysis step during RNA purification. 1 * 250 g
Catalog # ICNA0219400883
Supplier:  MP Biomedicals