Order Entry
Singapore
ContactUsLinkComponent
ISOPROPYL ALCOHOL. Molecular Biology ReagentPurity: 99+%Ideal for precipitating nucleic acids. In comparison to ethanol, approximate ly half the amount is required for precipitation which minimizes the total volume for recovery. 1 * 25 mL
  ICNA0219400625
ISOPROPYL ALCOHOL. Molecular Biology ReagentPurity: 99+%Ideal for precipitating nucleic acids. In comparison to ethanol, approximate ly half the amount is required for precipitation which minimizes the total volume for recovery. 1 * 25 mL
  ICNA0219400625