$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Cell Culture Reagent Anhydrous 1 * 500 g
ICNA0219474090
:
MP Biomedicals
Cell Culture Reagent Anhydrous 1 * 500 g
ICNA0219474090
:
MP Biomedicals