Order Entry
Singapore
ContactUsLinkComponent
RIBONUCLEASE A. From Bovine PancreasE.C. 2.7.7.16Molecular Biology ReagentChromatographically PurifiedLyophilized powderSalt-free an d protease-freeActivity: >70 Kunitz units/mgUnit Definition: One unit causes the hydrolysis of RNA at a rate such tha 1 * 50 mg
Catalog # ICNA0219398050
Supplier:  MP Biomedicals
RIBONUCLEASE A. From Bovine PancreasE.C. 2.7.7.16Molecular Biology ReagentChromatographically PurifiedLyophilized powderSalt-free an d protease-freeActivity: >70 Kunitz units/mgUnit Definition: One unit causes the hydrolysis of RNA at a rate such tha 1 * 50 mg
Catalog # ICNA0219398050
Supplier:  MP Biomedicals