Order Entry
Singapore
ContactUsLinkComponent
PANCREATIN 4X USP GRADE. From Porcine PancreasUSP Grade ReagentComposed of numerous enzymes including amylase, trypsin, lipase, ribo nuclease and protease. It will convert a minimum of 25 times its weight of potato starch into soluble carbohydrate in 1 * 500 g
Catalog # ICNA0219397590
Supplier:  MP Biomedicals
PANCREATIN 4X USP GRADE. From Porcine PancreasUSP Grade ReagentComposed of numerous enzymes including amylase, trypsin, lipase, ribo nuclease and protease. It will convert a minimum of 25 times its weight of potato starch into soluble carbohydrate in 1 * 500 g
Catalog # ICNA0219397590
Supplier:  MP Biomedicals