Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
LIDOCAINE HYDROCHLORIDE. Hydrochloride 1 * 25 g
Catalog # ICNA0219391725
Supplier:
MP Biomedicals
Expand image
LIDOCAINE HYDROCHLORIDE. Hydrochloride 1 * 25 g
Catalog # ICNA0219391725
Supplier:
MP Biomedicals