Order Entry
Singapore
ContactUsLinkComponent
INSULIN HUMAN RECOMBINANT. Human, Recombinant Expressed in E. coli Approx. 28 I.U. per mg Zinc content: 0.25 to 1.1% 1 * 100 mg
Catalog # ICNA0219390080
Supplier:  MP Biomedicals
INSULIN HUMAN RECOMBINANT. Human, Recombinant Expressed in E. coli Approx. 28 I.U. per mg Zinc content: 0.25 to 1.1% 1 * 100 mg
Catalog # ICNA0219390080
Supplier:  MP Biomedicals