Order Entry
Singapore
ContactUsLinkComponent
Purity: 99% Mixed Isomers Approx. 85% 1,3-isomer and 15% 1,2-isomer. Free of oleic acid. 1 * 1 g
Catalog # ICNA0219515191
Supplier:  MP Biomedicals
Purity: 99% Mixed Isomers Approx. 85% 1,3-isomer and 15% 1,2-isomer. Free of oleic acid. 1 * 1 g
Catalog # ICNA0219515191
Supplier:  MP Biomedicals