Order Entry
Singapore
ContactUsLinkComponent
LIMONIN. From Citrus SeedsA natural compound found in grapefruit and other citrus seeds. It has been shown to inhibit chemically induced carcinogenesis and demonstrates anti-feedant properties. 1 * 5 mg
Catalog # ICNA0219376505
Supplier:  MP Biomedicals
LIMONIN. From Citrus SeedsA natural compound found in grapefruit and other citrus seeds. It has been shown to inhibit chemically induced carcinogenesis and demonstrates anti-feedant properties. 1 * 5 mg
Catalog # ICNA0219376505
Supplier:  MP Biomedicals