Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
(Pentane-1,5-dial) 1 * 100 mL
Catalog # ICNA0219859580
Supplier:
MP Biomedicals
Expand image
(Pentane-1,5-dial) 1 * 100 mL
Catalog # ICNA0219859580
Supplier:
MP Biomedicals