Order Entry
Singapore
ContactUsLinkComponent
A general antiviral reported to be active against RSV, HIV, HCV, and other infectious agents. Its metabolite, ribavirin-5-phosphate, inhibits inosine monophosphate (IMP) dehydrogenase. 1 * 10 mg
Catalog # ICNA0219606610
Supplier:  MP Biomedicals
A general antiviral reported to be active against RSV, HIV, HCV, and other infectious agents. Its metabolite, ribavirin-5-phosphate, inhibits inosine monophosphate (IMP) dehydrogenase. 1 * 10 mg
Catalog # ICNA0219606610
Supplier:  MP Biomedicals