Order Entry
Singapore
ContactUsLinkComponent
TRICINE. (N-Tris-[hydroxymethyl]methylglycine) Cell Culture Reagent Biological buffer pKa at 25C = 8.1 Buffering pH range 7.4-8.8 1 * 100 g
Catalog # ICNA0219455680
Supplier:  MP Biomedicals
TRICINE. (N-Tris-[hydroxymethyl]methylglycine) Cell Culture Reagent Biological buffer pKa at 25C = 8.1 Buffering pH range 7.4-8.8 1 * 100 g
Catalog # ICNA0219455680
Supplier:  MP Biomedicals