Order Entry
Singapore
ContactUsLinkComponent
POLY-L-LYSINE HYDROBROMIDE gamma-IRRADIATED. Cell Culture Reagent-IrradiatedCellular attachment factor in culture applications. 1 * 5 mg
Catalog # ICNA0219454405
Supplier:  MP Biomedicals
POLY-L-LYSINE HYDROBROMIDE gamma-IRRADIATED. Cell Culture Reagent-IrradiatedCellular attachment factor in culture applications. 1 * 5 mg
Catalog # ICNA0219454405
Supplier:  MP Biomedicals