Order Entry
Singapore
ContactUsLinkComponent
Mouse, Purified, Monoclonal Clone: AE1/AE3 mixture Isotype: purified Ig fraction from ascites Conc/Titer: 1:100-1:200 Applications: Immunohistochemistry. 1 * 200 µG
Catalog # ICNA08691451
Supplier:  MP Biomedicals
Mouse, Purified, Monoclonal Clone: AE1/AE3 mixture Isotype: purified Ig fraction from ascites Conc/Titer: 1:100-1:200 Applications: Immunohistochemistry. 1 * 200 µG
Catalog # ICNA08691451
Supplier:  MP Biomedicals