Order Entry
Singapore
ContactUsLinkComponent
IgG Fc POLYCLONAL ANTIBODY. Anti-GoatHost: rabbitForm: affinity purified liquidApplications: ELISA, IEP and gel diffusion 1 * 1 mL
Catalog # ICNA0856021
Supplier:  MP Biomedicals
IgG Fc POLYCLONAL ANTIBODY. Anti-GoatHost: rabbitForm: affinity purified liquidApplications: ELISA, IEP and gel diffusion 1 * 1 mL
Catalog # ICNA0856021
Supplier:  MP Biomedicals