Order Entry
Singapore
Orders LinkContactUsLinkComponent
LiChrospher®, HPLC Columns
LiChrospher®, HPLC Columns
100Diol
Catalog # HICHLISPOH-5-250P
Supplier:   Avantor
LiChrospher®, HPLC Columns
Catalog # HICHLISPOH-5-250P
Supplier:  Avantor
LiChrospher®, HPLC Columns
LiChrospher®, HPLC Columns
100Diol
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
  • Spherical porous silica
  • Base deactivated LiChrospher® RP-select B
  • High efficiency packed columns
Specifications
  • Phase:
    100Diol