Order Entry
Singapore
ContactUsLinkComponent
GASG302 Built-In CO2 gas guard for Caron 7400, 7401, 7402, 7403 and 7404 series. Allows two CO2 tanks to be connected to a unit. Automatically switches from the primary tank to the secondary tank when low gas pressure is detected. Factory installed 1 * 1 items
Catalog # CARNGASG302
Supplier:  CARON PRODUCTS
GASG302 Built-In CO2 gas guard for Caron 7400, 7401, 7402, 7403 and 7404 series. Allows two CO2 tanks to be connected to a unit. Automatically switches from the primary tank to the secondary tank when low gas pressure is detected. Factory installed 1 * 1 items
Catalog # CARNGASG302
Supplier:  CARON PRODUCTS