Order Entry
Singapore
ContactUsLinkComponent
741-25-3 Carons 2-Tier, Shaker-Ready O2 Incubator is optimized for large-scale suspension cell culture applications using shakers. Standard features : Intuitive touchscreen controls, 2 shaker-support shelves, 3 internal outlets, IR CO2 sensor, and wi 1 * 1 items
Catalog # CARN7401-25-3
Supplier:  CARON PRODUCTS
741-25-3 Carons 2-Tier, Shaker-Ready O2 Incubator is optimized for large-scale suspension cell culture applications using shakers. Standard features : Intuitive touchscreen controls, 2 shaker-support shelves, 3 internal outlets, IR CO2 sensor, and wi 1 * 1 items
Catalog # CARN7401-25-3
Supplier:  CARON PRODUCTS