Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
CONDUCTIVITY STANDARD, 1000 µS/CM AT 25° 1 * 500 mL
RICCR2243000-500A
:
Ricca Chemical
CONDUCTIVITY STANDARD, 1000 µS/CM AT 25° 1 * 500 mL
RICCR2243000-500A
:
Ricca Chemical