Order Entry
Singapore
Orders LinkContactUsLinkComponent
 

Semi-micro balances, Explorer®

EX125, 120 g, 0,01 mg, Ø 80 mm, ±0,1 mg
Catalog # OHAU30139511
Supplier:   OHAUS
Semi-micro balances, Explorer®
Catalog # OHAU30139511
Supplier:  OHAUS
Semi-micro balances, Explorer®
Semi-micro balances, Explorer®

EX125, 120 g, 0,01 mg, Ø 80 mm, ±0,1 mg

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

These semi-microbalances have been designed and built for sophisticated professional weighing applications where the user requires extreme accuracy.

  • 'High Speed Single Module' weighing cell delivers ultra-fast stabilisation times typically ≤3 seconds for 0,1 mg and ≤8 seconds for 0,01 mg
  • Intuitive software with an icon-based colour touch screen display
  • AutoCal™ automatic internal calibration which uses two internal weights to perform a linear calibration
  • Save data to USB
  • Four touchless sensors allow the operator to perform common functions and commands with a swipe of the hand
  • Choice of anti-static glass draft shield with flip-top door, or Auto-Door - automatic draft shield doors for access to chamber without the need to touch the doors, minimising unwanted vibration
  • Built-in ioniser with Auto-Door models
  • LIMS compatibility

A user-manager function provides profiles for one administrator and five additional users together with a below minimum sample weight indicator and real time clock for GLP/GMP data collection.

 : A selection of OIML EC approved balances denoted M (e.g., EX125DM) are also available. For the full range of accessories for the Explorer® semi-micro balance please contact your local Avantor sales office.

Specifications
  • Model:
    EX125
  • Pan/Platform Size:
    Ø 80 mm
  • Seq:
    06
  • Readability:
    0,01 mg
  • Weighing cap.:
    120 g
  • Linearity:
    ±0,1 mg
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Density determination kit for Explorer®/Adventurer® balances (0,01,  0,1 and 1 mg models only)
    Density determination kit for Explorer®/Adventurer® balances (0,01,  0,1 and 1 mg models only)
    OHAU80253384
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Availability
    Price
  • Ethernet interface kit for Explorer® series
    Ethernet interface kit for Explorer® series
    OHAU83021082
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Availability
    Price
  • Sinker glass for density determination kit
    Sinker glass for density determination kit
    OHAU83034024
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Availability
    Price
  • Anti-theft device (Laptop lock) for Explorer®/Adventurer®/MB series
    Anti-theft device (Laptop lock) for Explorer®/Adventurer®/MB series
    OHAU80850043
    Log in for availability
    1 * 1 items