Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PHOSPHORIC ACID, 0.1% (V/V) AQUEOUS SOLU 1 * 4 L
Catalog # RICCR5845000-4A
Supplier:
Ricca Chemical
Expand image
PHOSPHORIC ACID, 0.1% (V/V) AQUEOUS SOLU 1 * 4 L
Catalog # RICCR5845000-4A
Supplier:
Ricca Chemical