Order Entry
Singapore
ContactUsLinkComponent
ACE® Excel Super C18 Ultra-Inert 3 and 5 micron Analytical UHPLC Compatible Columns, USP - L1, Mac Mod, Length 250mm 1 * 1 items
Catalog # VWRMNPEXL12112546U
Supplier:  VWR Products
ACE® Excel Super C18 Ultra-Inert 3 and 5 micron Analytical UHPLC Compatible Columns, USP - L1, Mac Mod, Length 250mm 1 * 1 items
Catalog # VWRMNPEXL12112546U
Supplier:  VWR Products