Buffers, pH reference standards, BDH®
Buffer, pH reference standard, 1,68, ±0,01, Colourless, 25 °C, Poly bottle, 4 L
Catalog # BDHB5008-4L
Supplier: BDH US PRODUCED
Jump to:
Product Details & Documents
Buffers are available as colourless and coloured solutions.
Certifications: ISO 17025 and traceable to NIST.
Documents
Specifications
Specifications
- Pack type:Poly bottle
- Pk:4 L
- Reference temperature:25 °C
- Colour:Colourless
- Description:Buffer, pH reference standard
- pH accuracy:±0,01
- pH value:1,68



