Order Entry
Singapore
Orders LinkContactUsLinkComponent
Buffers, pH reference standards, BDH®

Buffers, pH reference standards, BDH®

Buffer, pH reference standard, 10,00, ±0,01, Blue, 25 °C, Cubitainer®, 20 L
Catalog # BDHB0190-20L
Supplier:   BDH US PRODUCED
Buffers, pH reference standards, BDH®
Catalog # BDHB0190-20L
Supplier:  BDH US PRODUCED
CAS Number:  
Buffers, pH reference standards, BDH®

Buffer, pH reference standard, 10,00, ±0,01, Blue, 25 °C, Cubitainer®, 20 L

CAS Number:  
Price
$713.00
Log in for availability
1 * 20 L
Product Details & Documents

Buffers are available as colourless and coloured solutions.

 : ISO 17025 and traceable to NIST.

Specifications

Specifications

  • Pack type:
    Cubitainer®
  • Pk:
    20 L
  • Reference temperature:
    25 °C
  • Colour:
    Blue
  • Description:
    Buffer, pH reference standard
  • pH accuracy:
    ±0,01
  • pH value:
    10,00

Specifications