Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER PH 4.01 2.5GAL 2.5GL 1 * 1 items
Catalog # RICCR1501000-10F
Supplier:
Ricca Chemical
Expand image
BUFFER PH 4.01 2.5GAL 2.5GL 1 * 1 items
Catalog # RICCR1501000-10F
Supplier:
Ricca Chemical