Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Flask Conical, borosilicate glass 3.3 250ml, socket 24/29 1 * 1 items
Catalog # IBSAF90004-0019C
Supplier:
IBS Stock Items
Expand image
Flask Conical, borosilicate glass 3.3 250ml, socket 24/29 1 * 1 items
Catalog # IBSAF90004-0019C
Supplier:
IBS Stock Items