NucleoMag Kits for Pathogen Detection
NucleoMag VET kit for viral/bacterial DNA and viral RNA from veterinary samples, 2032,7 g, 384 preps, can be used for PCR, enzymatic reactions, RT-PCR, Isolation of viral and bacterial nucleic acids, Magnetic beads, Animal tissue, Plasma, Serum, Blood, Stool, Swabs, Cell-free biological fluid, 50 - 100 µl
Catalog # MANA744200.4
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
NucleoMag kits are designed for rapid manual and automated small-scale preparation of viral RNA and DNA and the DNA of microorganisms from various types of clinical samples. NucleoMag® kits allow easy automation on common liquid handling instruments or automated magnetic separators.
- Magnetic bead technology
- NucleoMag® kits for pathogen detection are designed for the isolation of viral RNA and DNA and bacterial DNA from cell-free body fluids such as serum or plasma, blood, saliva, homogenized tissue sample suspensions, stool sample suspensions, and swab washes
Support protocols for automation platforms available on request.
Handling: Magnetic separation.
Hazardous material: No.
Automation support: Compatible with most open robotic platforms.
: Shelf life: 24 months from date of production. For research use only.
Specifications
Specifications
- Description:NucleoMag VET kit for viral/bacterial DNA and viral RNA from veterinary samples, 2032,7 g, 384 preps, can be used for PCR, enzymatic reactions, RT-PCR, Isolation of viral and bacterial nucleic acids
- Format:Magnetic beads
- Volume:50 - 100 µl
- Contents:Animal tissue, Plasma, Serum, Blood, Stool, Swabs, Cell-free biological fluid
- Cat. No.:MANA744200.4
- Prep size:Scalable
- Refine target molecule:Viral RNA and DNA
- Target molecule:Copurification of DNA and RNA
- Method / Format:Magnetic beads
- Binding capacity:0,4 µg/µl beads
- Operation time:40 - 120 min/96 preps
- Sample size:<200 µl whole blood/serum/plasma/swab solution/feces, 10 - 30 mg tissue
- Fragment length:300 bp - approx. 50 kbp
- Storage temperature:15...25 °C (59...77 °F)
- Recovery:Depending on sample amount and quality



