Order Entry
Singapore
Orders LinkContactUsLinkComponent
NucleoMag Kits for Pathogen Detection

NucleoMag Kits for Pathogen Detection

NucleoMag VET kit for viral/bacterial DNA and viral RNA from veterinary samples, 625 g, 96 preps, can be used for PCR, enzymatic reactions, RT-PCR, Isolation of viral and bacterial nucleic acids, Magnetic beads, Animal tissue, Plasma, Serum, Blood, Stool, Swabs, Cell-free biological fluid, 50 - 100 µl
Catalog # MANA744200.1
Supplier:   MACHEREY-NAGEL
NucleoMag Kits for Pathogen Detection
Catalog # MANA744200.1
Supplier:  MACHEREY-NAGEL
CAS Number:  
NucleoMag Kits for Pathogen Detection
NucleoMag Kits for Pathogen Detection

NucleoMag VET kit for viral/bacterial DNA and viral RNA from veterinary samples, 625 g, 96 preps, can be used for PCR, enzymatic reactions, RT-PCR, Isolation of viral and bacterial nucleic acids, Magnetic beads, Animal tissue, Plasma, Serum, Blood, Stool, Swabs, Cell-free biological fluid, 50 - 100 µl

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 96 Tests
Product Details & Documents

NucleoMag kits are designed for rapid manual and automated small-scale preparation of viral RNA and DNA and the DNA of microorganisms from various types of clinical samples. NucleoMag® kits allow easy automation on common liquid handling instruments or automated magnetic separators.

  • Magnetic bead technology
  • NucleoMag® kits for pathogen detection are designed for the isolation of viral RNA and DNA and bacterial DNA from cell-free body fluids such as serum or plasma, blood, saliva, homogenized tissue sample suspensions, stool sample suspensions, and swab washes

Support protocols for automation platforms available on request.

Handling: Magnetic separation.

Hazardous material: No.

Automation support: Compatible with most open robotic platforms.

Caution: Shelf life: 24 months from date of production. For research use only.

Specifications

Specifications

  • Description:
    NucleoMag VET kit for viral/bacterial DNA and viral RNA from veterinary samples, 625 g, 96 preps, can be used for PCR, enzymatic reactions, RT-PCR, Isolation of viral and bacterial nucleic acids
  • Format:
    Magnetic beads
  • Volume:
    50 - 100 µl
  • Contents:
    Animal tissue, Plasma, Serum, Blood, Stool, Swabs, Cell-free biological fluid
  • Cat. No.:
    MANA744200.1
  • Prep size:
    Scalable
  • Refine target molecule:
    Viral RNA and DNA
  • Target molecule:
    Copurification of DNA and RNA
  • Method / Format:
    Magnetic beads
  • Binding capacity:
    0,4 µg/µl beads
  • Operation time:
    40 - 120 min/96 preps
  • Sample size:
    <200 µl whole blood/serum/plasma/swab solution/feces, 10 - 30 mg tissue
  • Fragment length:
    300 bp - approx. 50 kbp
  • Storage temperature:
    15...25 °C (59...77 °F)
  • Recovery:
    Depending on sample amount and quality

Specifications