Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Electrophoresis VS20 WAVE Maxi System, 20 x 20cm Dual with Glass plates, 2 x 24 well combs, cooling coil (No casting Base) 1 * 1 items
CLEVVS20WAVED
:
CLEAVER SCIENTIFIC
Electrophoresis VS20 WAVE Maxi System, 20 x 20cm Dual with Glass plates, 2 x 24 well combs, cooling coil (No casting Base) 1 * 1 items
CLEVVS20WAVED
:
CLEAVER SCIENTIFIC