Accessories for data loggers, Dickson®
Software and USB cable for data loggers, DicksonWare™ SECURE
Catalog # DICKA026
Supplier: DICKSON
Jump to:
Product Details & Documents
DicksonWare™ software (incl. USB or serial cable) is needed for setting up Dickson loggers. They enable viewing and downloading data, and resetting the logger to the factory settings.
Specifications
- Description:Software and USB cable for data loggers, DicksonWare™ SECURE
- Cat. No.:DICKA026
- Interface:USB