Accessories for data loggers, Dickson®
Software and USB cable for data loggers, DicksonWare™ SECURE
DICKA025
: DICKSON
DicksonWare™ software (incl. USB or serial cable) is needed for setting up Dickson loggers. They enable viewing and downloading data, and resetting the logger to the factory settings.
- Description:Software and USB cable for data loggers, DicksonWare™ SECURE