Order Entry
Singapore
ContactUsLinkComponent
Plug-in tube extension (outer diam. 60,3 mm, inner 57 mm), straight, for exhaust air ducting (if necessary for connection by hose), only models U, I, S 1 * 1 items
Catalog # MEMMB29718UF1060
Supplier:  MEMMERT
Plug-in tube extension (outer diam. 60,3 mm, inner 57 mm), straight, for exhaust air ducting (if necessary for connection by hose), only models U, I, S 1 * 1 items
Catalog # MEMMB29718UF1060
Supplier:  MEMMERT