Order Entry
Singapore
ContactUsLinkComponent
Custom bottle for Roche - 2 L FEP Bottle with cap, pre-cleaned Materials supplied by EP Process: USP Purified Rinse & Particulate Cleaning Note - Outside of the bottles washed first with caps engaged in order to avoid rinsing particles from the outer 1 * 2 items
  NALG7M2000WP-EP
Custom bottle for Roche - 2 L FEP Bottle with cap, pre-cleaned Materials supplied by EP Process: USP Purified Rinse & Particulate Cleaning Note - Outside of the bottles washed first with caps engaged in order to avoid rinsing particles from the outer 1 * 2 items
  NALG7M2000WP-EP