Biotix 1250ul Pipette Tips. Racked, 10 racks of 96 Tips/Pack, 4 Packs/Case. Universal fit graduated, Patented features: FlexFit for universal fit and superior seal, Chem-Resin for drip-free pipetting of polar solvents, and Blade for accuracy 1 * 960 items
Biotix 1250ul Pipette Tips. Racked, 10 racks of 96 Tips/Pack, 4 Packs/Case. Universal fit graduated, Patented features: FlexFit for universal fit and superior seal, Chem-Resin for drip-free pipetting of polar solvents, and Blade for accuracy 1 * 960 items