Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Stirrer Blades, PTFE, 10mm OD, Blade Width 19 mm, Length 48 mm 1 * 1 items
Catalog # ACEG8085-03
Supplier:
Ace Glass
Expand image
Stirrer Blades, PTFE, 10mm OD, Blade Width 19 mm, Length 48 mm 1 * 1 items
Catalog # ACEG8085-03
Supplier:
Ace Glass